Product Details
- SNP ID
-
rs199663847
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:105659205 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCTGCGCGGTCGTGGTTTAGGG[G/T]GTCTGGCTGGTGCCTGTGAACCATC
- Phenotype
-
MIM: 604491
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CBLB
PubMed Links
Gene Details
- Gene
- CBLB
- Gene Name
- Cbl proto-oncogene B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321786.1 |
2600 |
Missense Mutation |
CAC,CCC |
H933P |
NP_001308715.1 |
NM_001321788.1 |
2600 |
Missense Mutation |
CAC,CCC |
H905P |
NP_001308717.1 |
NM_001321789.1 |
2600 |
Missense Mutation |
CAC,CCC |
H884P |
NP_001308718.1 |
NM_001321790.1 |
2600 |
Missense Mutation |
CAC,CCC |
H883P |
NP_001308719.1 |
NM_001321791.1 |
2600 |
Missense Mutation |
CAC,CCC |
H861P |
NP_001308720.1 |
NM_001321793.1 |
2600 |
Missense Mutation |
CAC,CCC |
H861P |
NP_001308722.1 |
NM_001321794.1 |
2600 |
Missense Mutation |
CAC,CCC |
H856P |
NP_001308723.1 |
NM_001321795.1 |
2600 |
Missense Mutation |
CAC,CCC |
H856P |
NP_001308724.1 |
NM_001321796.1 |
2600 |
Missense Mutation |
CAC,CCC |
H856P |
NP_001308725.1 |
NM_001321797.1 |
2600 |
Missense Mutation |
CAC,CCC |
H812P |
NP_001308726.1 |
NM_001321798.1 |
2600 |
Missense Mutation |
CAC,CCC |
H812P |
NP_001308727.1 |
NM_001321799.1 |
2600 |
Missense Mutation |
CAC,CCC |
H812P |
NP_001308728.1 |
NM_001321806.1 |
2600 |
Missense Mutation |
CAC,CCC |
H645P |
NP_001308735.1 |
NM_001321807.1 |
2600 |
Missense Mutation |
CAC,CCC |
H645P |
NP_001308736.1 |
NM_001321808.1 |
2600 |
Missense Mutation |
CAC,CCC |
H601P |
NP_001308737.1 |
NM_001321811.1 |
2600 |
Missense Mutation |
CAC,CCC |
H596P |
NP_001308740.1 |
NM_001321813.1 |
2600 |
Missense Mutation |
CAC,CCC |
H596P |
NP_001308742.1 |
NM_001321816.1 |
2600 |
Missense Mutation |
CAC,CCC |
H552P |
NP_001308745.1 |
NM_001321820.1 |
2600 |
Missense Mutation |
CAC,CCC |
H465P |
NP_001308749.1 |
NM_001321822.1 |
2600 |
Missense Mutation |
CAC,CCC |
H462P |
NP_001308751.1 |
NM_170662.4 |
2600 |
Missense Mutation |
CAC,CCC |
H905P |
NP_733762.2 |
XM_011513257.1 |
2600 |
Missense Mutation |
CAC,CCC |
H905P |
XP_011511559.1 |
XM_011513259.2 |
2600 |
Intron |
|
|
XP_011511561.2 |
XM_017007395.1 |
2600 |
Missense Mutation |
CAC,CCC |
H927P |
XP_016862884.1 |
XM_017007396.1 |
2600 |
Missense Mutation |
CAC,CCC |
H889P |
XP_016862885.1 |
XM_017007397.1 |
2600 |
Missense Mutation |
CAC,CCC |
H884P |
XP_016862886.1 |
XM_017007398.1 |
2600 |
Missense Mutation |
CAC,CCC |
H878P |
XP_016862887.1 |
XM_017007399.1 |
2600 |
Missense Mutation |
CAC,CCC |
H856P |
XP_016862888.1 |
XM_017007400.1 |
2600 |
Missense Mutation |
CAC,CCC |
H840P |
XP_016862889.1 |
View Full Product Details