Product Details

SNP ID
rs200997202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99885571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTGTTATAGTAACAGATGATAC[C/T]TTTATTTTTGGAAAGTCCAAGTTTG
Phenotype
MIM: 603296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR3 PubMed Links

Gene Details

Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243736.1 457 Missense Mutation AGT,GGT S63G NP_001230665.1
NM_021970.3 457 Missense Mutation AGT,GGT S70G NP_068805.1

View Full Product Details