Product Details

SNP ID
rs200746802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4267892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTTTTAAATACACAAATGTTCA[C/T]TTCACAAGCTTGACTTTGTCCTTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYAR PubMed Links

Gene Details

Gene
LYAR
Gene Name
Ly1 antibody reactive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145725.1 1311 Silent Mutation AAA,AAG K379K NP_001139197.1
NM_017816.2 1311 Silent Mutation AAA,AAG K379K NP_060286.1
XM_011513505.1 1311 Silent Mutation AAA,AAG K379K XP_011511807.1
XM_011513506.2 1311 Silent Mutation AAA,AAG K379K XP_011511808.1

View Full Product Details