Product Details

SNP ID
rs200192631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186533709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTTAAACGGAGTCCACCTTTA[C/T]TACATTATTGTTGGTCATCAGTGGA
Phenotype
MIM: 600665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTNR1A PubMed Links

Gene Details

Gene
MTNR1A
Gene Name
melatonin receptor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005958.4 1235 Missense Mutation ATA,GTA I345V NP_005949.1
XM_011532002.2 1235 Missense Mutation ATA,GTA I260V XP_011530304.1

View Full Product Details