Product Details

SNP ID
rs199702688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102885295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTGCAGTTCGTCCATTCCTCCC[A/G]AAGAAGAAACAACAGAAGGATAGCT
Phenotype
MIM: 611507 MIM: 611527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CISD2 PubMed Links

Gene Details

Gene
CISD2
Gene Name
CDGSH iron sulfur domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008388.4 1508 Silent Mutation CCA,CCG P61P NP_001008389.1
Gene
SLC9B1
Gene Name
solute carrier family 9 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100874.2 1508 Missense Mutation CGG,TGG R456W NP_001094344.1
NM_139173.3 1508 Intron NP_631912.2
XM_006714093.3 1508 Intron XP_006714156.1
XM_011531622.1 1508 Intron XP_011529924.1
XM_011531623.1 1508 Intron XP_011529925.1

View Full Product Details