Product Details

SNP ID
rs199796507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7041722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAATCTTCAGCTGTTCAAAGTCA[A/G]TAAGAAGCAGACCCTGGGTCAGGTC
Phenotype
MIM: 608790 MIM: 614855
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC96 PubMed Links

Gene Details

Gene
CCDC96
Gene Name
coiled-coil domain containing 96
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153376.2 1280 Missense Mutation NP_699207.1
Gene
LOC100129931
Gene Name
uncharacterized LOC100129931
There are no transcripts associated with this gene.

Gene
TADA2B
Gene Name
transcriptional adaptor 2B
There are no transcripts associated with this gene.

Gene
TBC1D14
Gene Name
TBC1 domain family member 14
There are no transcripts associated with this gene.

View Full Product Details