Product Details

SNP ID
rs199837068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69486673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGAGTCCTAAAGTATCTGGTTT[A/G]TTCCCATCAAATCTCCACAGAACCT
Phenotype
MIM: 600067
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGT2B4 PubMed Links

Gene Details

Gene
UGT2B4
Gene Name
UDP glucuronosyltransferase family 2 member B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297615.1 1073 Silent Mutation AAC,AAT N342N NP_001284544.1
NM_001297616.1 1073 Silent Mutation AAC,AAT N206N NP_001284545.1
NM_021139.2 1073 Silent Mutation AAC,AAT N342N NP_066962.2

View Full Product Details