Product Details

SNP ID
rs199769280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55080091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGTGTCATCGGAGTGATATCC[A/G]GACTGGTAGCCGCTTGTCTGGTTTG
Phenotype
MIM: 191306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDR PubMed Links

Gene Details

Gene
KDR
Gene Name
kinase insert domain receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002253.2 4223 Silent Mutation TCC,TCT S1307S NP_002244.1

View Full Product Details