Product Details

SNP ID
rs202015819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:11399414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGCAGCCAGTTCTGCATGTGCA[C/T]GTGGTAGAGGCTGCGGTTGAGGGCC
Phenotype
MIM: 603244
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HS3ST1 PubMed Links

Gene Details

Gene
HS3ST1
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005114.2 5674 Missense Mutation ATG,GTG M198V NP_005105.1
XM_011513913.2 5674 Missense Mutation ATG,GTG M198V XP_011512215.1

View Full Product Details