Product Details

SNP ID
rs201931730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70689135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGAGGCCGGCAGAGTCAACAGG[A/C]GGCAGAGGAGGAGGAAAGAGAGGAG
Phenotype
MIM: 611614
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UTP3 PubMed Links

Gene Details

Gene
UTP3
Gene Name
UTP3, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020368.2 657 Missense Mutation GAG,GCG E153A NP_065101.1

View Full Product Details