Product Details
- SNP ID
-
rs202121184
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:3767078 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACTGGTCGGTGACGCAGGCCGTC[A/G]AGTACAACCTGAAGCGCACACCACG
- Phenotype
-
MIM: 104250
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADRA2C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138457504] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADRA2C
- Gene Name
- adrenoceptor alpha 2C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000683.3 |
510 |
Missense Mutation |
AAG,GAG |
K158E |
NP_000674.2 |
View Full Product Details