Product Details

SNP ID
rs202238726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:164957278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAGAAGCGAATCGTTTTCTCC[C/G]CGTGCCCTGTCAGCCGCTCATGGTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM218A PubMed Links

Gene Details

Gene
FAM218A
Gene Name
family with sequence similarity 218 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153027.1 331 Missense Mutation NP_694572.1
Gene
TRIM61
Gene Name
tripartite motif containing 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012414.2 331 Intron NP_001012414.1

View Full Product Details