Product Details

SNP ID
rs201984942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154477497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTCCGCTTGCTCTCACGGCCG[C/T]CATCCGCCTTCTTGAGTTCAGCCTT
Phenotype
MIM: 602406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAND1 PubMed Links

Gene Details

Gene
HAND1
Gene Name
heart and neural crest derivatives expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004821.2 713 Missense Mutation GAC,GGC D171G NP_004812.1
XM_005268531.1 713 Missense Mutation GAC,GGC D171G XP_005268588.1

View Full Product Details