Product Details

SNP ID
rs202220094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:24488103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATATCTTCTTTTGACAAGATCA[A/G]AGGCTCTTTTTTTCGCTGTCTTTTC
Phenotype
MIM: 604555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH10 PubMed Links

Gene Details

Gene
CDH10
Gene Name
cadherin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317222.1 1930 Silent Mutation CTG,TTG L49L NP_001304151.1
NM_001317224.1 1930 Silent Mutation CTG,TTG L641L NP_001304153.1
NM_006727.4 1930 Silent Mutation CTG,TTG L643L NP_006718.2
XM_011513923.2 1930 Silent Mutation CTG,TTG L643L XP_011512225.1
XM_017008915.1 1930 Silent Mutation CTG,TTG L641L XP_016864404.1

View Full Product Details