Product Details
- SNP ID
-
rs201913253
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:111092420 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGACTGGGTACGTGTTTTCCTTC[A/G]GGACCAGAGCTGAGAGGAGCTGGGA
- Phenotype
-
MIM: 609669
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR36
PubMed Links
Gene Details
- Gene
- WDR36
- Gene Name
- WD repeat domain 36
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139281.2 |
249 |
Silent Mutation |
TCA,TCG |
S44S |
NP_644810.1 |
View Full Product Details