Product Details

SNP ID
rs202056255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150691794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCATGAGCTCCCATCCTCTGA[C/G]GGTCCTGAGAAGGATAGTGGATTGG
Phenotype
MIM: 612430
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBM22 PubMed Links

Gene Details

Gene
RBM22
Gene Name
RNA binding motif protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018047.2 1342 Missense Mutation CCT,CGT P407R NP_060517.1

View Full Product Details