Product Details

SNP ID
rs199737472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40681374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCATCAACCATGCCTATTTCTA[C/T]AGCCACTACGTGGACAAGCGATTGG
Phenotype
MIM: 601586
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGER4 PubMed Links

Gene Details

Gene
PTGER4
Gene Name
prostaglandin E receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000958.2 973 Silent Mutation TAC,TAT Y127Y NP_000949.1
XM_017009656.1 973 Silent Mutation TAC,TAT Y127Y XP_016865145.1
XM_017009657.1 973 Silent Mutation TAC,TAT Y127Y XP_016865146.1
XM_017009658.1 973 Silent Mutation TAC,TAT Y127Y XP_016865147.1
XM_017009659.1 973 Silent Mutation TAC,TAT Y127Y XP_016865148.1

View Full Product Details