Product Details

SNP ID
rs200157804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141628119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAACTGACAGTATTACCTGGA[C/G]CACAATGCACGTGGGTTGGTAGAAG
Phenotype
MIM: 602121 MIM: 605166 MIM: 611213
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DIAPH1 PubMed Links

Gene Details

Gene
DIAPH1
Gene Name
diaphanous related formin 1
There are no transcripts associated with this gene.

Gene
HDAC3
Gene Name
histone deacetylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003883.3 826 Missense Mutation CTC,GTC L254V NP_003874.2
XM_011537697.2 826 Missense Mutation CTC,GTC L67V XP_011535999.1
Gene
RELL2
Gene Name
RELT like 2
There are no transcripts associated with this gene.

View Full Product Details