Product Details

SNP ID
rs199604249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154820359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTCTGGGATGCTCTCAGAGAGC[C/G]GGGTGAAGCCCAGCAGGAGGACATG
Phenotype
MIM: 612059
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAXDC2 PubMed Links

Gene Details

Gene
FAXDC2
Gene Name
fatty acid hydroxylase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032385.4 807 Missense Mutation CCG,CGG P320R NP_115761.2
XM_005268358.2 807 Missense Mutation CCG,CGG P180R XP_005268415.1
XM_006714753.2 807 Missense Mutation CCG,CGG P320R XP_006714816.1
XM_017008965.1 807 Missense Mutation CCG,CGG P320R XP_016864454.1
XM_017008966.1 807 Missense Mutation CCG,CGG P180R XP_016864455.1
Gene
LARP1
Gene Name
La ribonucleoprotein domain family member 1
There are no transcripts associated with this gene.

Gene
MIR378H
Gene Name
microRNA 378h
There are no transcripts associated with this gene.

View Full Product Details