Product Details

SNP ID
rs199921823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141215416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGACTTGGGGACCCCTATGCT[A/G]ATAACACAGCTCAATATGACCGTGC
Phenotype
MIM: 606338 MIM: 606339 MIM: 606340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB12 PubMed Links

Gene Details

Gene
PCDHB12
Gene Name
protocadherin beta 12
There are no transcripts associated with this gene.

Gene
PCDHB13
Gene Name
protocadherin beta 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018933.3 1498 Silent Mutation CTA,CTG L431L NP_061756.1
Gene
PCDHB14
Gene Name
protocadherin beta 14
There are no transcripts associated with this gene.

View Full Product Details