Product Details

SNP ID
rs200282241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9629767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCAAGCCAAAGTTCCAATTCA[C/T]TTATAAATAAGAGAATTGCACAATT
Phenotype
MIM: 604796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R1 PubMed Links

Gene Details

Gene
TAS2R1
Gene Name
taste 2 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019599.2 585 Missense Mutation AAT,AGT N89S NP_062545.1

View Full Product Details