Product Details

SNP ID
rs201139003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:205713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTGCATCACCTGGGCCCGTT[A/C]CTGTTCCAGAAGCTTCTTTTCTTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC127 PubMed Links

Gene Details

Gene
CCDC127
Gene Name
coiled-coil domain containing 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145265.2 467 Nonsense Mutation GAA,TAA E123* NP_660308.1

View Full Product Details