Product Details

SNP ID
rs201139557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:143820541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGATGATGTGTATCTGAGGCA[C/T]AGCTCTTCCCTGACTTATAGGCTTT
Phenotype
MIM: 609961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMHB1 PubMed Links

Gene Details

Gene
HMHB1
Gene Name
histocompatibility minor HB-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021182.1 206 Silent Mutation CAC,CAT H33H NP_067005.1

View Full Product Details