Product Details

SNP ID
rs201104078
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:24487922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGACGTCCGTGTTATCTGGAGCT[G/T]TAGGAGTCCTCCGAGGAATAAATAA
Phenotype
MIM: 604555
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CDH10 PubMed Links

Gene Details

Gene
CDH10
Gene Name
cadherin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317222.1 2111 Missense Mutation AGA,ATA R109I NP_001304151.1
NM_001317224.1 2111 Missense Mutation AGA,ATA R701I NP_001304153.1
NM_006727.4 2111 Missense Mutation AGA,ATA R703I NP_006718.2
XM_011513923.2 2111 Missense Mutation AGA,ATA R703I XP_011512225.1
XM_017008915.1 2111 Missense Mutation AGA,ATA R701I XP_016864404.1

View Full Product Details