Product Details

SNP ID
rs201575174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177386235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCATCCAGAGTCCAGGCTGGTC[C/G]CCAAGCTGCGCCAGGCTGGCGCCAT
Phenotype
MIM: 182309
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC34A1 PubMed Links

Gene Details

Gene
SLC34A1
Gene Name
solute carrier family 34 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167579.1 321 Missense Mutation CCC,GCC P92A NP_001161051.1
NM_003052.4 321 Missense Mutation CCC,GCC P92A NP_003043.3
XM_005265975.1 321 Missense Mutation CCC,GCC P92A XP_005266032.1
XM_017009773.1 321 Missense Mutation CCC,GCC P92A XP_016865262.1
XM_017009774.1 321 Intron XP_016865263.1
XM_017009775.1 321 Missense Mutation CCC,GCC P92A XP_016865264.1

View Full Product Details