Product Details

SNP ID
rs201087525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141351221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAGTGTGGAAGCTAAGGATGGA[A/G]GAGTACACACAGCTCACTGTAATGT
Phenotype
MIM: 606288 MIM: 606289 MIM: 606290 MIM: 606291 MIM: 606299 MIM: 606300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 961 Intron NP_061735.1
NM_031993.1 961 Intron NP_114382.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018915.3 961 Intron NP_061738.1
NM_032009.2 961 Intron NP_114398.1
Gene
PCDHGA3
Gene Name
protocadherin gamma subfamily A, 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018916.3 961 Intron NP_061739.2
NM_032011.1 961 Intron NP_114400.1
Gene
PCDHGA4
Gene Name
protocadherin gamma subfamily A, 4
There are no transcripts associated with this gene.

Gene
PCDHGB1
Gene Name
protocadherin gamma subfamily B, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018922.2 961 Missense Mutation AGA,GGA R321G NP_061745.1
NM_032095.1 961 Missense Mutation AGA,GGA R321G NP_115266.1
Gene
PCDHGB2
Gene Name
protocadherin gamma subfamily B, 2
There are no transcripts associated with this gene.

View Full Product Details