Product Details

SNP ID
rs201030012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55256515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTATTCTCTGTTTTTATCAATTC[C/T]GTAATGATCTGCAGAATCTTGTCAT
Phenotype
MIM: 612720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX29 PubMed Links

Gene Details

Gene
DHX29
Gene Name
DEAH-box helicase 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019030.2 4040 Silent Mutation ACA,ACG T1361T NP_061903.2
XM_006714653.2 4040 Silent Mutation ACA,ACG T1310T XP_006714716.1
XM_017009588.1 4040 Silent Mutation ACA,ACG T725T XP_016865077.1

View Full Product Details