Product Details
- SNP ID
-
rs200793188
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:16451760 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCATCCTTTGGACATACTGCATG[C/T]CTCGCTCTCGCATAAGAGCCGCTCC
- Phenotype
-
MIM: 608694
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF622
PubMed Links
Gene Details
- Gene
- ZNF622
- Gene Name
- zinc finger protein 622
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033414.2 |
1451 |
Missense Mutation |
GAC,GGC |
D444G |
NP_219482.1 |
View Full Product Details