Product Details

SNP ID
rs201606826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10227561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTCCCCCGTGACGTGGTCTGG[A/G]GTCCAATGTGGGAAAGGGAACCGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM173B PubMed Links

Gene Details

Gene
FAM173B
Gene Name
family with sequence similarity 173 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258388.1 621 Silent Mutation ACC,ACT T177T NP_001245317.1
NM_001258389.1 621 UTR 3 NP_001245318.1
NM_199133.3 621 Silent Mutation ACC,ACT T194T NP_954584.2
XM_011513964.1 621 UTR 3 XP_011512266.1

View Full Product Details