Product Details
- SNP ID
-
rs201943873
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26251962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACAGCCGTGCGCCTGCTGCTGCC[G/T]GGGGAACTGGCCAAGCACGCCGTGT
- Phenotype
-
MIM: 602806
MIM: 602816
MIM: 602832
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HIST1H2BH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61746493] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIST1H2BH
- Gene Name
- histone cluster 1, H2bh
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003524.2 |
|
Intron |
|
|
NP_003515.1 |
- Gene
- HIST1H3F
- Gene Name
- histone cluster 1, H3f
There are no transcripts associated with this gene.
- Gene
- HIST1H4G
- Gene Name
- histone cluster 1, H4g
There are no transcripts associated with this gene.
View Full Product Details