Product Details

SNP ID
rs202123656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87085823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTTCTCCACTTTGAAACCCCCC[A/G]TTACTGTGACCTAAAGGGGAAGGAA
Phenotype
MIM: 118850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CGA PubMed Links

Gene Details

Gene
CGA
Gene Name
glycoprotein hormones, alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000735.3 519 Missense Mutation ACG,ATG T95M NP_000726.1
NM_001252383.1 519 Missense Mutation ACG,ATG T126M NP_001239312.1

View Full Product Details