Product Details

SNP ID
rs201855901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18121742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACTTTCACCCTGGTGCTGCAAA[C/G]CCCAGTCCCCAGGGCCAGGGGGTGC
Phenotype
MIM: 608072 MIM: 187680
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NHLRC1 PubMed Links

Gene Details

Gene
NHLRC1
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198586.2 879 Missense Mutation CTT,GTT L289V NP_940988.2
Gene
TPMT
Gene Name
thiopurine S-methyltransferase
There are no transcripts associated with this gene.

View Full Product Details