Product Details

SNP ID
rs201016286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41030670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGTCTCACCTATTGCATGGGGGC[A/G]GCTGGGAAACAGGTGGTGGCGCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929555 PubMed Links

Gene Details

Gene
LOC101929555
Gene Name
uncharacterized LOC101929555
There are no transcripts associated with this gene.

Gene
UNC5CL
Gene Name
unc-5 family C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173561.2 1304 Missense Mutation CCG,CTG P402L NP_775832.2

View Full Product Details