Product Details

SNP ID
rs200483949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118966900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTTAATTCTGTAATCATCTGTA[C/T]ATCTTCTGGTTTTGATTCTCTCATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM184A PubMed Links

Gene Details

Gene
FAM184A
Gene Name
family with sequence similarity 184 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100411.2 3316 Missense Mutation ATA,GTA I821V NP_001093881.1
NM_001288576.1 3316 Missense Mutation ATA,GTA I821V NP_001275505.1
NM_024581.5 3316 Missense Mutation ATA,GTA I990V NP_078857.5

View Full Product Details