Product Details

SNP ID
rs200449082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116647502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGAGGTTTAGATGTACACACTAC[C/T]TTTGGACTCCCTTCTCCCTCTGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZUFSP PubMed Links

Gene Details

Gene
ZUFSP
Gene Name
zinc finger with UFM1 specific peptidase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145062.2 2078 Silent Mutation AAA,AAG K475K NP_659499.2
XM_011535565.2 2078 Silent Mutation AAA,AAG K475K XP_011533867.1
XM_011535567.1 2078 Silent Mutation AAA,AAG K307K XP_011533869.1
XM_017010393.1 2078 Silent Mutation AAA,AAG K475K XP_016865882.1
XM_017010394.1 2078 Silent Mutation AAA,AAG K279K XP_016865883.1
XM_017010395.1 2078 Silent Mutation AAA,AAG K251K XP_016865884.1
XM_017010396.1 2078 Silent Mutation AAA,AAG K251K XP_016865885.1

View Full Product Details