Product Details

SNP ID
rs200886860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17601091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGTAGGATCCGCAGCCCCTTCG[C/G]GAAGCCCGAGCGAGACCGGGCGACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM8A1 PubMed Links

Gene Details

Gene
FAM8A1
Gene Name
family with sequence similarity 8 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016255.2 805 Missense Mutation CGA,GGA R228G NP_057339.1

View Full Product Details