Product Details

SNP ID
rs200868601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116635847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGATTCTTCAAGGAATCTTCTC[A/G]GCTGTAAAGACTTGAGAAGCTTGTC
Phenotype
MIM: 612647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RSPH4A PubMed Links

Gene Details

Gene
RSPH4A
Gene Name
radial spoke head 4 homolog A
There are no transcripts associated with this gene.

Gene
ZUFSP
Gene Name
zinc finger with UFM1 specific peptidase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145062.2 2375 Silent Mutation GCC,GCT A574A NP_659499.2
XM_011535565.2 2375 Silent Mutation GCC,GCT A574A XP_011533867.1
XM_011535567.1 2375 Silent Mutation GCC,GCT A406A XP_011533869.1
XM_017010393.1 2375 Intron XP_016865882.1
XM_017010394.1 2375 Silent Mutation GCC,GCT A378A XP_016865883.1
XM_017010395.1 2375 Silent Mutation GCC,GCT A350A XP_016865884.1
XM_017010396.1 2375 Silent Mutation GCC,GCT A350A XP_016865885.1

View Full Product Details