Product Details

SNP ID
rs201533442
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27452189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGTTGAGCAAGGGATGACCAG[A/T]AACTAAAAGATTTTCCACATTCTGC
Phenotype
MIM: 602277
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 1655 Missense Mutation TAC,TTC Y457F NP_001305820.1
NM_001318892.1 1655 Missense Mutation TAC,TTC Y457F NP_001305821.1
NM_001318893.1 1655 Missense Mutation TAC,TTC Y381F NP_001305822.1
NM_007149.2 1655 Missense Mutation TAC,TTC Y457F NP_009080.2
XM_011514861.2 1655 Missense Mutation TAC,TTC Y381F XP_011513163.1

View Full Product Details