Product Details

SNP ID
rs201502336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31575764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGAGCACTGAAAGCATGATCC[A/G]GGACGTGGAGCTGGCCGAGGAGGCG
Phenotype
MIM: 153440 MIM: 600978 MIM: 191160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100287329 PubMed Links

Gene Details

Gene
LOC100287329
Gene Name
uncharacterized LOC100287329
There are no transcripts associated with this gene.

Gene
LTA
Gene Name
lymphotoxin alpha
There are no transcripts associated with this gene.

Gene
LTB
Gene Name
lymphotoxin beta
There are no transcripts associated with this gene.

Gene
TNF
Gene Name
tumor necrosis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000594.3 198 Missense Mutation CAG,CGG Q8R NP_000585.2

View Full Product Details