Product Details

SNP ID
rs201349903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149969350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGATGTACTGGGAACAAATGCT[C/G]GATCCAACAAGTAAGTGAGAGGGGG
Phenotype
MIM: 605697
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ULBP1 PubMed Links

Gene Details

Gene
ULBP1
Gene Name
UL16 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317089.1 691 Intron NP_001304018.1
NM_025218.3 691 Intron NP_079494.1
XM_017011322.1 691 Silent Mutation CTC,CTG L205L XP_016866811.1

View Full Product Details