Product Details

SNP ID
rs199797213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27871917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATAGGAATATTTATGCCCTCTC[C/G]CCTCGGATGCGGCGCGCAAGCTGGA
Phenotype
MIM: 142711 MIM: 602793 MIM: 602814 MIM: 602831
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H1B PubMed Links

Gene Details

Gene
HIST1H1B
Gene Name
histone cluster 1, H1b
There are no transcripts associated with this gene.

Gene
HIST1H2AL
Gene Name
histone cluster 1, H2al
There are no transcripts associated with this gene.

Gene
HIST1H3I
Gene Name
histone cluster 1, H3i
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003533.2 Intron NP_003524.1
Gene
HIST1H4L
Gene Name
histone cluster 1, H4l
There are no transcripts associated with this gene.

View Full Product Details