Product Details

SNP ID
rs200172498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49786227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACAACTTGAGCATGATTCTTGT[G/T]GCTCTCAGGACCACAGTCCAAACCC
Phenotype
MIM: 172270
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PGK2 PubMed Links

Gene Details

Gene
PGK2
Gene Name
phosphoglycerate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138733.4 1114 Missense Mutation AAC,CAC N321H NP_620061.2

View Full Product Details