Product Details

SNP ID
rs200055869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:112071523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGATGACACAGCTTCTGTGAA[A/G]CAGCTTTCTTCCATCCCTTCAACTT
Phenotype
MIM: 607345 MIM: 603400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TUBE1 PubMed Links

Gene Details

Gene
TUBE1
Gene Name
tubulin epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016262.4 1445 Silent Mutation TGC,TGT C439C NP_057346.1
XM_011535875.2 1445 Silent Mutation TGC,TGT C364C XP_011534177.1
XM_017010918.1 1445 Silent Mutation TGC,TGT C395C XP_016866407.1
Gene
WISP3
Gene Name
WNT1 inducible signaling pathway protein 3
There are no transcripts associated with this gene.

View Full Product Details