Product Details

SNP ID
rs199622510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82173372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTATCAATTAACCTTACTTGGG[A/G]CTGTCTAGAAGTGGTAAATCTGAAA
Phenotype
MIM: 606457
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IBTK PubMed Links

Gene Details

Gene
IBTK
Gene Name
inhibitor of Bruton tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300906.1 5005 Silent Mutation AGC,AGT S1249S NP_001287835.1
NM_015525.3 5005 Silent Mutation AGC,AGT S1264S NP_056340.2
XM_006715453.2 5005 Silent Mutation AGC,AGT S1305S XP_006715516.1
XM_006715454.2 5005 Silent Mutation AGC,AGT S1290S XP_006715517.1

View Full Product Details