Product Details

SNP ID
rs199644346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55174678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACTCAAGAGCCCTTTTTAAAC[C/T]CCACCGACTATGACGACGAGGAATT
Phenotype
MIM: 602393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCRTR2 PubMed Links

Gene Details

Gene
HCRTR2
Gene Name
hypocretin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001526.3 406 Missense Mutation CCC,TCC P31S NP_001517.2
XM_017010798.1 406 Missense Mutation CCC,TCC P31S XP_016866287.1

View Full Product Details