Product Details

SNP ID
rs200215521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGGGAAGCTGCGCGGACGAGGA[A/G]GAGGTCCCCGAGGAGTGGGTGCTCC
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 447 Silent Mutation GAA,GAG E33E NP_001025034.1
NM_001317342.1 447 Silent Mutation GAA,GAG E52E NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 447 Intron NP_001189479.1
NM_182552.4 447 Intron NP_872358.4
XM_011535682.2 447 Intron XP_011533984.1
XM_011535684.2 447 Intron XP_011533986.1
XM_011535685.2 447 Intron XP_011533987.1
XM_011535687.2 447 Intron XP_011533989.1
XM_011535688.2 447 Intron XP_011533990.1
XM_011535689.2 447 Intron XP_011533991.1
XM_011535690.2 447 Intron XP_011533992.1
XM_011535691.2 447 Intron XP_011533993.1
XM_011535692.2 447 Intron XP_011533994.1
XM_011535693.2 447 Intron XP_011533995.1
XM_011535694.2 447 Intron XP_011533996.1
XM_011535696.2 447 Intron XP_011533998.1
XM_011535697.2 447 Intron XP_011533999.1
XM_017010658.1 447 Intron XP_016866147.1
XM_017010659.1 447 Intron XP_016866148.1
XM_017010660.1 447 Intron XP_016866149.1
XM_017010661.1 447 Intron XP_016866150.1
XM_017010662.1 447 Intron XP_016866151.1
XM_017010663.1 447 Intron XP_016866152.1
XM_017010664.1 447 Intron XP_016866153.1
XM_017010665.1 447 Intron XP_016866154.1
XM_017010666.1 447 Intron XP_016866155.1
XM_017010667.1 447 Intron XP_016866156.1
XM_017010668.1 447 Intron XP_016866157.1
XM_017010669.1 447 Intron XP_016866158.1
XM_017010670.1 447 Intron XP_016866159.1
XM_017010671.1 447 Intron XP_016866160.1
XM_017010672.1 447 Intron XP_016866161.1
XM_017010673.1 447 Intron XP_016866162.1
XM_017010674.1 447 Intron XP_016866163.1
XM_017010675.1 447 Intron XP_016866164.1

View Full Product Details