Product Details

SNP ID
rs200742026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5622926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGTGCTCCAGGGGCATGTGGAT[A/G]GGACCGAAGTCATAGTTGACCCGCA
Phenotype
MIM: 609948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107986762 PubMed Links

Gene Details

Gene
LOC107986762
Gene Name
uncharacterized LOC107986762
There are no transcripts associated with this gene.

Gene
RNF216
Gene Name
ring finger protein 216
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207111.3 2704 Silent Mutation CCC,CCT P902P NP_996994.1
NM_207116.2 2704 Silent Mutation CCC,CCT P845P NP_996999.1
XM_005249785.2 2704 Silent Mutation CCC,CCT P902P XP_005249842.1
XM_011515436.1 2704 Silent Mutation CCC,CCT P467P XP_011513738.1
XM_017012363.1 2704 Silent Mutation CCC,CCT P845P XP_016867852.1
XM_017012364.1 2704 Intron XP_016867853.1
XM_017012365.1 2704 Silent Mutation CCC,CCT P467P XP_016867854.1

View Full Product Details