Product Details

SNP ID
rs200817164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142908755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCCTTGTCTGAGTTCTTGAAC[A/T]CTTCCACATAGCGAAGCACTCGCAG
Phenotype
MIM: 606679
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TRPV5 PubMed Links

Gene Details

Gene
TRPV5
Gene Name
transient receptor potential cation channel subfamily V member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019841.6 2213 Missense Mutation GAG,GTG E650V NP_062815.3

View Full Product Details