Product Details

SNP ID
rs199951557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6409860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAGGCCACGTCCACACTGGAGA[C/T]CCCTTGTGCTGGACAGGAGACGCAG
Phenotype
MIM: 614016 MIM: 602048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAGLB PubMed Links

Gene Details

Gene
DAGLB
Gene Name
diacylglycerol lipase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142936.1 2166 Missense Mutation ATC,GTC I537V NP_001136408.1
NM_139179.3 2166 Missense Mutation ATC,GTC I666V NP_631918.3
Gene
RAC1
Gene Name
ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
There are no transcripts associated with this gene.

View Full Product Details