Product Details

SNP ID
rs200671062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141935826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGTTCTCACCAGTTTCTCTGG[C/T]GTGTTGACAATTGCCAATGTGGATC
Phenotype
MIM: 604987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLEC5A PubMed Links

Gene Details

Gene
CLEC5A
Gene Name
C-type lectin domain family 5 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301167.1 417 Silent Mutation ACA,ACG T88T NP_001288096.1
NM_013252.2 417 Silent Mutation ACA,ACG T111T NP_037384.1
XM_011515995.2 417 Silent Mutation ACA,ACG T68T XP_011514297.1
XM_017011915.1 417 Silent Mutation ACA,ACG T91T XP_016867404.1
XM_017011916.1 417 Silent Mutation ACA,ACG T111T XP_016867405.1
XM_017011917.1 417 Silent Mutation ACA,ACG T88T XP_016867406.1

View Full Product Details